Learn More
Cayman Chemical FurIn InhIbItr ItrIfluoroa
SDP
Supplier:  Cayman Chemical 14965500 UG
Encompass_Preferred
A selective, irreversible, and cell-permeable competitive inhibitor of proprotein convertases, including furin/SPC1 (Ki = ~1 nM), SPC2/PC2 (Ki = 0.36 nM), SPC3/PC1/PC3 (Ki = 2.0 nM), SPC4/PACE4 (Ki = 3.6 nM), SPC6/PC5/PC6, and SPC7/LPC/PC7/PC8 (Ki = 0.12 nM)
Catalog No. 50-297-0685