Learn More
Yamato Scientific America, Inc. Yamato 3.75L Standard Muffle Furnace with Communication Port, 115V
Supplier: Yamato Scientific America, Inc. FO200CR
Standard 3.75L muffle furnace using microprocessor-controlled thermo-regulator characterized by excellent repeatability. Used for high temperature applications such as fusing glass, creating enamel coatings, ceramics and soldering and brazing metals. Easy to use controller. Excellent heat tightness with a firmly sealed chamber door. High temperature accuracy at ±2.0°C. Program operation of maximum of 6 patterns: 30 steps x 1 pattern, 15 steps x 2 patterns or 10 steps x 3 patterns. Safety features include self-diagnostic functions, calibration off-set, lock function, auto-recovery after power failure, earth leakage breaker and automatic overheat prevention device. Selectable options include exhaust system unit, N₂ gas loading device (with flow meter), temperature output terminal, time up/alarm output terminal and sample tray. Upgraded with long life R-thermocouple sensors. Designed with communication port. Temperature range: 100~1150°C.