Learn More
Medchemexpress LLC Furosemide | 54-31-9 | 100.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0135R50MG
Encompass_Preferred
Furosemide is an analytical standard intended for research and analytical applications. It is a potent, orally active inhibitor of Na+/K+/2Cl- (NKCC) cotransporter (NKCC1 and NKCC2) and a GABAA receptor antagonist, showing 100-fold selectivity for α6-containing receptors over α1-containing receptors. Furosemide functions as a loop diuretic and is used for studying congestive heart failure, hypertension, and edema. It is commonly utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Potent and orally active inhibitor of NKCC cotransporter
  • GABAA receptor antagonist with high selectivity for α6-containing receptors
  • Functions as a loop diuretic
  • Used for studying congestive heart failure, hypertension, and edema
  • Suitable for qualitative, quantitative, and methodological research experiments
  • Applicable in HPLC, GC, and MS analyses
Catalog No. 50-004-50468