Learn More
Medchemexpress LLC Furosemide (sodium) | 41733-55-5 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0135A10MM/1ML
Encompass_Preferred
Furosemide sodium is a potent and orally active inhibitor of Na+/K+/2Cl- (NKCC) cotransporter, specifically NKCC1 and NKCC2. It also acts as a GABAA receptors antagonist with 100-fold selectivity for α6-containing receptors over α1-containing receptors. This compound functions as a loop diuretic and is utilized in research for conditions such as congestive heart failure, hypertension, and edema.
  • Inhibitor of NKCC cotransporter (NKCC1 and NKCC2)
  • GABAA receptors antagonist
  • Selective for α6-containing receptors
  • Functions as a loop diuretic
  • Used in research for congestive heart failure
  • Used in research for hypertension
  • Used in research for edema
Catalog No. 50-004-50885