Learn More
Medchemexpress LLC Furosemide (standard) | 54-31-9 | 99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0135R100MG
Encompass_Preferred
Furosemide (Standard) is an analytical reference standard for research and analytical applications. It acts as a potent and orally active inhibitor of Na+/K+/2Cl- (NKCC) cotransporter, specifically NKCC1 and NKCC2, and also functions as a GABAA receptor antagonist. This compound is used in the study of congestive heart failure, hypertension, and edema, and is commonly employed in qualitative, quantitative, and methodological research experiments like HPLC, GC, and MS.
  • Potent and orally active inhibitor of Na+/K+/2Cl- (NKCC) cotransporter
  • Functions as a GABAA receptor antagonist
  • Exhibits 100-fold selectivity for α6-containing receptors over α1-containing receptors
  • Acts as a loop diuretic
  • Utilized in the study of congestive heart failure, hypertension, and edema
  • Analytical standard grade
  • Suitable for qualitative, quantitative, and methodological research experiments
Catalog No. 50-004-50509