Learn More
Cayman Chemical Fusarenon X 100ug
SDP
Supplier:  Cayman Chemical 11432100 UG
Encompass_Preferred
A type B trichothecene mycotoxin found in cereals contaminated with certain Fusarium species
Catalog No. 50-295-2106