Learn More
Medchemexpress LLC HY-15009 5mg Medchemexpress, Fuscoside CAS:131631-89-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY150095MG
Encompass_Preferred
Medchemexpress, HY-15009 5mg Fuscoside CAS:131631-89-5 Fuscoside (OPC-21268) is an orally effective, nonpeptide, vasopressin V1 receptor antagonist with an IC50 of 0.4 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1670