Learn More
Grainger Fuse Block 31 to 60A 600VAC
Supplier:  Grainger 14L611
Encompass_Preferred
The BUSSMANN Finger-Safe Fuse Block with Cover is a 30A, 600VAC finger-safe fuse block that has screw terminals and accepts 1/4"x1-1/4" fuses. It features a clear cover for safety and visibility. This UL listed block can be panel, surface, or DIN rail mounted for versatility. It has a high impact resistant polycarbonate cover and melamine base that is rated for both indoor and outdoor use. The cover features a tamper-resistant design to prevent unauthorized fuse access. This is an ideal finger-safe fuse block for protecting circuits and equipment.
Catalog No. 50-274-9701