Learn More
Grainger Fuse Block 31 to 60A 600VAC
Supplier:  Grainger 14L612
Encompass_Preferred
The Bussmann Finger-Safe Fuse Block with Cover is a fuse block that provides overcurrent protection for electrical circuits. It has space for up to 6 ATC/ATO automotive blade fuses and features a clear plastic hinged cover for safety. The maximum voltage is 32V DC and maximum current is 60A. The terminals accept 10-22 AWG wire. It can be mounted via the included screw or attached to a surface with adhesive. This finger-safe fuse block is ideal for protecting electrical circuits in vehicles, RVs, boats, and more.
Catalog No. 50-274-8708