Learn More
NEWARK ELEMENT14 FUSE, CARTRIDGE, 6.3A, 5X20MM
Supplier:  NEWARK ELEMENT14 88K2034
Encompass
FUSE, CARTRIDGE, 6.3A, 5X20MM TIME DELAY; Voltage Rating VAC:250V; Fuse Current:6.3A; Breaking Capacity Current AC:1.5kA; Fuse Size Metric:5mm x 20mm; Fuse Size Imperial:0.2 in. x 0.79 in.; Blow Characteristic:Time Delay; Series:SPT
Catalog No. NC0416155