Learn More
Medchemexpress LLC Fusicoccin | 20108-30-9 | 96.3% | 680.82 | 1 MG
SDP
Supplier:  Medchemexpress LLC 1MGHY122815
Encompass_Preferred
Fusicoccin is a fungal phytotoxin that stabilizes specific 14-3-3 protein-protein interactions, including the H+-ATPase/14-3-3 complex in plants. It also stabilizes 14-3-3 protein interactions with binding partners containing a C-terminal 14-3-3 recognition motif. This compound induces apoptosis in cancer cells and exhibits anticancer activity.
  • Stabilizes specific 14-3-3 protein-protein interactions
  • Induces apoptosis in cancer cells
  • Exhibits anticancer activity
  • Activates plasma membrane H+-ATPase in plants
  • Decreases proliferation and migration of human GBM cell lines in vitro
Catalog No. 50-003-65389