Learn More
Chemplex Industries FUSION FLUXES PRE-FUSED 80%
Supplier:  Chemplex Industries 303000
Encompass_Preferred
CHEMPLEX FUSION FLUXES; PRE-FUSED; 1 lb/btl (454 gm), 80% Li-Tetra./20% Li-Meta.
Catalog No. 50-249-542