Learn More
TARGETMOL CHEMICALS INC FUTIBATINIB 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T50445MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Futibatinib (FGFR-IN-1) is a novel potent and highly selective FGFR inhibitor used for antitumor treatment. purity: 99%
Catalog No. 50-257-6711