Learn More
Selleck Chemical LLC Futibatinib S8848-10MM/1ML
SDP
Supplier:  Selleck Chemical LLC S884810MM/1ML
Encompass_Preferred
Futibatinib (TAS-120) is an irreversible fibroblast growth factor receptor (FGFR) inhibitor which inhibits all 4 subtypes of FGFR with enzyme IC50 values of 1 8 nM 1 4 nM 1 6 nM and 3 7 nM for FGFR1 FGFR2 FGFR3 and FGFR4 respectively
Catalog No. 50-000-24774