Learn More
TARGETMOL CHEMICALS INC FV-100 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T411165MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. FV-100 (Valnivudine HCl) is a potent selective and orally active anti-vaxx-zoster agent.FV-100 is the API for CF-1743.FV-100 exhibits very low toxicity in vivo.FV-100 is a potent selective and orally active anti-vaxx-zoster agent.FV-100 is the API for CF-1743.FV-100 exhibits very low toxicity in vivo. purity: 99%
Catalog No. 50-257-9959