Learn More
Medchemexpress LLC FXIIa-IN-4 | 98.9% | 264.21 | 1 ML

Supplier: Medchemexpress LLC HY16334810MM/1ML
FXIIa-IN-4 is a potent and selective human Factor XIIa (FXIIa) inhibitor with an IC50 of 0.032 μM. It exhibits significantly lower inhibitory activity against thrombin (IC50 of 0.30 μM) and FXIa (IC50 > 50 μM), demonstrating its selectivity. This compound is primarily used for research purposes to study anticoagulation.
- Potent and selective human FXIIa inhibitor.
- Exhibits high selectivity for FXIIa over thrombin and FXIa.
- Suitable for anticoagulation research.
- High purity of 98.87%.
- Available in various quantities, including solid and solution forms.
- Detailed documentation available, including data sheet, COA, and SDS.
- Chemical formula is C11H9FN4O3 with a molecular weight of 264.21.
- Solid appearance, white to off-white in color.
- Soluble in DMSO at 25 mg/mL.
- Can be stored long-term as powder at -20°C for 3 years, or in solvent at -80°C for 6 months.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.