Learn More
Aviva Systems Biology Corp FZD7 Antibody - C-terminal region (ARP41251_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41251_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FZD7. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human FZD7. Alternate names: FzE3ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-57781