Learn More
Medchemexpress LLC FzM1 100mg | 1680196-54-6 | 100 MG

Supplier: Medchemexpress LLC HY116553100MG
FzM1 is a small-molecule negative allosteric modulator (NAM) of the Frizzled receptor FZD4 that reduces WNT5A-dependent WNT responsive element (WRE) activity and inhibits WNT/β-catenin signaling. It binds an allosteric site in intracellular loop 3 (ICL3) of FZD4 and is supplied as a solid research reagent.
- Negative allosteric modulator of the FZD4 receptor.
- Reduces WNT5A-dependent WNT responsive element (WRE) activity (log EC50inh ≈ -6.2).
- Molecular formula C21H16N2O2S; molecular weight 360.43.
- Purity approximately 98.7% as supplied.
- Storage: powder at -20°C; in solvent -80°C up to 6 months or -20°C up to 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.