Learn More
Selleck Chemical LLC G-1 S0851-5MG
SDP
Supplier:  Selleck Chemical LLC S08515MG
Encompass_Preferred
G-1 (LNS 8801) is a nonsteroidal and selective agonist of G protein-coupled receptor 30 (GPR30 G protein-coupled estrogen receptor 1 GPER) with Ki of 11 nM
Catalog No. 50-000-42935