Learn More
MESA LABS BOZEMAN MFG FACILITY G. 10 6 SPORE STRIPS 100/BAG
SDP
Supplier:  MESA LABS BOZEMAN MFG FACILITY S2X10/6G
Encompass
2x10mm 10e6 micro-strips for steam sterilization - Glassine-packaged
Catalog No. NC9815718