Learn More
Cayman Chemical G-15, 1161002-05-6, 5 mg
SDP
Supplier:  Cayman Chemical 146735
Encompass_Preferred
Molecular Formula C19H16BrNO2, Purity ≥95%, Formula Weight 370.2; A cell-permeable non-steroidal antagonist of the estrogen receptor GPER (Ki = 20 nM); displays low affinity cross-reactivity with the classical estrogen receptor, ER, so that at doses greater than 1 M it is capable of mediating limited ER-dependent transcriptional activity.
Catalog No. NC1343291