Learn More
Cayman Chemical G-36, 1392487-51-2, 5 mg
SDP
Supplier:  Cayman Chemical 143975
Encompass_Preferred
Molecular Formula C22H22BrNO2, Purity ≥98%, Formula Weight 412.3; A cell-permeable non-steroidal antagonist of GPER, inhibiting activation by either 17-estradiol or the GPER-selective agonist G-1 (IC50 = 112 and 165 nM, respectively); has no detectable binding activity to either ER or ER.
Catalog No. NC1081973