Learn More
Alkali Scientific G 418 Disulfate 50 Mg/mL Solution, 10 Milliliters
SDP
Supplier:  Alkali Scientific GDI005CP
Encompass_Preferred
G 418 Disulfate 50 mg/mL solution (10mL) is a highly effective antibiotic solution used in cell culture selection. This solution allows for the selection of cells that have successfully incorporated the neomycin resistance gene. The 50 mg/mL concentration in this 10mL vial makes it ideal for researchers working with mammalian cells in genetic modification experiments. The liquid form of G418 offers convenience and ease of use, ensuring precise application without the need for further dilution. It is widely used in gene expression studies, transfection research, and genetic knockout studies.
Catalog No. 50-006-61012