Learn More
AG Scientific Inc G-418.Sulfate, 50G
Supplier:  AG Scientific Inc G103350GM
Encompass_Preferred
G-418 Sulfate Salt (Geneticin) is an aminoglycoside antibiotic that is synthesized from Micromonospora rhodorangea and functions by causing a chain of nonsense mutations during translation. Due to translation G-418 inhibited polypeptide synthesis and protein elongation. G-418 binds to the 80S subunit of the ribosome by blocking the elongation phase of protein synthesis, both in prokaryotic and eukaryotic cells. It also shows activity against protozoa, cecal amoebiasis, and helminthes.CAS Number: 108321-42-2Molecular Weight: 692.7 DaChemical Formula: C20H40O10N4 2H2SO4Solubility: Soluble in water: 100mg/ml, colorless solutionStorage Temperature: +4CResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-6915