Learn More
Medchemexpress LLC G-CSF Mouse CHO 500ug
SDP
Supplier:  Medchemexpress LLC HYP7068500UG
Encompass_Preferred
Recombinant mouse granulocyte colony-stimulating factor (G-CSF) produced in CHO cells, provided for research applications to regulate neutrophil production, differentiation, and function. Commonly used in studies of hematopoiesis, inflammation, and immune response, the product is supplied in a laboratory-scale pack suitable for assay development and experimental workflows.
  • Recombinant protein expressed in CHO cells for mammalian-like post-translational modifications.
  • Regulates neutrophil proliferation and differentiation for hematopoietic studies.
  • Suitable for in vitro and in vivo research applications.
  • Provided in a laboratory-scale pack size for experimental use.
  • Compatible with standard cell culture and biochemical assays.
Catalog No. 50-001-84303