Learn More
Medchemexpress LLC G-CSF Rat HEK293 2ug
SDP
Supplier:  Medchemexpress LLC HYP7093A2UG
Encompass_Preferred
G-CSF protein (rat) expressed in HEK293 cells is a glycosylated recombinant cytokine that stimulates granulopoiesis, supports innate immune responses, and promotes differentiation of neural progenitor cells. The lyophilized material is supplied for in vitro biological assays and activity testing, with documented high specific activity and low endotoxin suitable for cell-based experiments.
  • Recombinant rat G-CSF produced in HEK293 cells.
  • Glycosylated protein with observed molecular weight ~27-32 kDa.
  • Stimulates granulopoiesis and supports innate immunity.
  • Promotes differentiation of neural progenitor cells.
  • High biological potency (ED50 ≤ 16.25 pg/mL; specific activity ≥ 6.15 x 10^7 U/mg).
  • Low endotoxin level (<1 EU/μg) for cell-based assays.
  • Lyophilized formulation; store frozen and use aliquots after reconstitution.
  • Reconstitute at ≥100 μg/mL in ddH2O for recommended handling.
Catalog No. 50-001-97147