Learn More
Medchemexpress LLC (-)-G-Lactone | 43119-28-4 | ≥98.0% | 124.14 | 500 MG

Supplier: Medchemexpress LLC HY43001500MG
(-)-G-Lactone is a synthetic intermediate useful for prostaglandins synthesis. It is for research use only and not sold to patients.
- Appears as a solid
- Color is off-white to light yellow
- Store at room temperature for 3 years
- Store in solvent at -80°C for 2 years or -20°C for 1 year
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.