Learn More
CPC Scientific H-Gly-Arg-Gly-Asp-Ser-Pro-Cys-OH (trifluoroacetate salt) 25MG
SDP
Supplier:  CPC Scientific FIBN015C
Encompass
ONE-LETTER SEQUENCE: GRGDSPCMOLECULAR FORMULA: C25H42N10O11S1MOLECULAR WEIGHT:690.73STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [126646-79-5], SYNONYMS: Gly-Arg-Gly-Asp-Ser-Pro-Cys, GRGDSPCRESEARCH AREA: InflammationREFERENCES: M.D. Pieschbacher et al., PNAS, 81, 5985 (1984)
Catalog No. 50-210-9298