Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC G0-C14 | 1510653-27-6 | C106H216N10O10 | 1 ML
SDP

Supplier:  Medchemexpress LLC HY15222910MM/1ML

Encompass_Preferred

G0-C14 is a cationic lipid-like compound and an alkyl-modified polyamidoamine (PAMAM) dendrimer. It is utilized in the preparation of macrophage-targeted nanoparticles (NPs), which are effective for agent and vaccine delivery. This compound demonstrates strong entrapment of both mRNA and pDNA, achieving over 95% encapsulation efficiency.

  • Cationic lipid-like compound.
  • Alkyl-modified polyamidoamine (PAMAM) dendrimer.
  • Used for preparation of macrophage-targeted nanoparticles.
  • Supports agent and vaccine delivery.
  • Strong entrapment of mRNA and pDNA.
  • Over 95% encapsulation efficiency.
  • Colorless to light yellow oil appearance.
  • Purity greater than 98.0%.

Catalog No. 50-003-72089


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.