Learn More
Medchemexpress LLC HY-128583 10mg , G150 CAS:2369751-30-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12858310MG
Encompass_Preferred
HY-128583 10mg G150 CAS: 2369751-30-2 G150 is a highly selective human cyclic GMP-AMP synthase (h-cGAS) inhibitor with an IC50 of 10.2 nM. G150 represses dsDNA-triggered interferon expression, and G150 can be used for the research of inflammatory. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0020