Learn More
Grainger G2228 6-Inch Work Boot M 10 Brown PR
Supplier:  Grainger 16P474
Encompass_Preferred
The FisherbrandTM 502768654 is a polypropylene microcentrifuge tube rack that holds 0.5mL tubes. It has 96 holes arranged in an 8x12 matrix to accommodate up to 96 tubes. The rack is made from autoclavable polypropylene and can withstand temperatures from -20°C to 121°C. It has alphanumeric grid references for easy sample identification. The overall dimensions are 127mm x 76mm x 45mm. This microcentrifuge tube rack provides an organized storage solution for 0.5mL tubes in research and clinical settings.
Catalog No. 50-276-6113