Learn More
Grainger G2386 Arc Flash Body Harness ExoFit NEX S
Supplier:  Grainger 16V456
Encompass_Preferred
The 3M DBI-SALA Full Body Harness Arc Flash is a full body harness designed for fall protection and arc flash protection. It features a lightweight, breathable design with a back D-ring for fall arrest and shoulder, chest, and leg D-rings for positioning. The arc-rated webbing provides protection against electric arc flash hazards. This harness has a weight capacity of 130-420 lbs and comes in sizes ranging from small to 4XL. It meets OSHA, ANSI, and ASTM standards for fall protection and arc flash safety. The harness is made with flame-resistant materials including Nomex webbing.
Catalog No. 50-276-5040