Learn More
Medchemexpress LLC G3-C12 (TFA) | 99.4% | 1873.00 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP1592A10MG
Encompass_Preferred
G3-C12 (TFA) is a galectin-3 binding peptide that exhibits anticancer activity with a Kd of 88 nM. It demonstrates no affinity for other galectin family members or other lectins. When bearing opolymers N-(2-hydroxypropyl)methacrylamide (HPMA), it potently and selectively targets colorectal cancer (CRC) tumor cells over-expressing galectin-3.
  • Galectin-3 binding peptide
  • Exhibits anticancer activity
  • Has a Kd of 88 nM
  • Shows no affinity for other galectin family members or other lectins
  • Potently and selectively targets colorectal cancer (CRC) tumor cells over-expressing galectin-3
Catalog No. 50-004-22171