Learn More
Invivogen G418 5 G (100 MG/ML)
SDP
Catalog No. NC9227938
Encompass
G418, also known as G418 sulfate and Geneticin, is an aminoglycoside antibiotic similar in structure to gentamicin B1, produced by Micromonospora rhodorangea. G418 blocks polypeptide synthesis by inhibiting the elongation step in both prokaryotic and eukaryotic cells. Resistance to G418 is conferred by the Neomycin resistance gene (neo) from Tn5 encoding an aminoglycoside 3‘-phosphotransferase, APH 3‘ II. Selection in mammalian cells is usually achieved in three to seven days with concentrations ranging from 400 to 1000 µg/ml. Cells that are dividing are affected sooner than those that are not.
Catalog No. NC9227938 Supplier Invivogen Supplier No. ANTGN5
G418, also known as G418 sulfate and Geneticin, is an aminoglycoside antibiotic similar in structure to gentamicin B1, produced by Micromonospora rhodorangea. G418 blocks polypeptide synthesis by inhibiting the elongation step in both prokaryotic and eukaryotic cells. Resistance to G418 is conferred by the Neomycin resistance gene (neo) from Tn5 encoding an aminoglycoside 3‘-phosphotransferase, APH 3‘ II. Selection in mammalian cells is usually achieved in three to seven days with concentrations ranging from 400 to 1000 µg/ml. Cells that are dividing are affected sooner than those that are not.