Learn More
Cell Signaling Technology G4S LINKER(E7O2V)RAB,MAB,100UL
SDP
Supplier:  Cell Signaling Technology 38907S
Encompass
G4S Linker (E7O2V) Rabbit mAb (PE Conjugate) 100 ulThis Cell Signaling Technology antibody is conjugated to phycoerythrin (PE) fluorescent dye under optimal conditions and tested in-house for direct flow cytometric analysis in human cells. This antibody conjugate is expected to exhibit the same reactivity as the unconjugated G4S Linker (E7O2V) Rabbit mAb #71645, which is expected to react with cell surface-expressed CARs of varying specificity containing a G4S linker within the scFv of the extracellular domain.
Catalog No. NC2292152