Learn More
TARGETMOL CHEMICALS INC G6PDI-1 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T864010MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. G6PDi-1 is an effective G6PD inhibitor. It depletes NADPH and decreases inflammatory cytokine production. purity: 98%
Catalog No. 50-257-6233