Learn More
Medchemexpress LLC GA32 | 1386811-71-7 | 99.7% | 417.48 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1633405MG
Encompass_Preferred
GA32 (compound 58r) is a potent androgen receptor (AR)/glucocorticoid receptor (GR) dual inhibitor with IC50 values of 0.13 μM and 0.83 μM for AR and GR, respectively. GA32 inhibits the proliferation of Enzalutamide resistance castration-resistant prostate cancer both in vitro and in vivo.
  • Potent androgen receptor (AR)/glucocorticoid receptor (GR) dual inhibitor.
  • Inhibits proliferation of Enzalutamide resistance castration-resistant prostate cancer in vitro and in vivo.
Catalog No. 50-003-92509