Learn More
ABclonal Technology GAB1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2349920UL
Encompass_Preferred
The protein encoded by this gene is a member of the IRS1-like multisubstrate docking protein family It is an important mediator of branching tubulogenesis and plays a central role in cellular growth response transformation and apoptosis Two transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-06038