Learn More
Medchemexpress LLC GABA B Receptor 2 Antibody (YA1733) | 10 UL
SDP
Supplier:  Medchemexpress LLC HYP8198810UL
Encompass_Preferred
GABA B Receptor 2 Antibody (YA1733) is a Rabbit-derived, non-conjugated IgG monoclonal antibody targeting GABA B Receptor 2. It is for research use only and reacts with Human, Mouse, and Rat species. It is supplied as a liquid in 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40%Glycerol, 0.01% sodium azide and 0.05% BSA.
  • Rabbit-derived and non-conjugated IgG monoclonal antibody
  • Targets GABA B Receptor 2
  • Reactivity with Human, Mouse, Rat
  • Available in multiple sizes
  • Endogenous sensitivity
  • Affinity Chromatography purity
  • Unmodified
Catalog No. 50-004-41657