Learn More
Enzo Life Sciences Gabapentin (100mg). CAS: 60142-96-3
SDP
Supplier:  Enzo Life Sciences BMLCA2340100
Encompass_Preferred
Gabapentin is a GABA analog which does not act at GABA receptors but which inhibits L-type calcium channels via binding to the α2-δ subunit and neuropathic pain relieving actions. Ligands acting at the α2-δ subunit lead to anticonvulsant, anxiolytic and neuropathic pain-relieving actions. Purity: ≥98% (HPLC). Solubility: Soluble in water. Long Term Storage: Ambient.
Catalog No. 50-201-0374