Learn More
TARGETMOL CHEMICALS INC Gabapentin 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T0702200MG
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Gabapentin (Neurontin) is an Anti-epileptic Agent. The physiologic effect of gabapentin is by means of Decreased Central Nervous System Disorganized Electrical Activity. Purity 99.39%
Catalog No. 50-288-0635