Learn More
Medchemexpress LLC Gabapentin enacarbil | 478296-72-9 | 95.0% | 329.39 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1621610MM/1ML
Encompass_Preferred
Gabapentin enacarbil (XP-13512) is a prodrug of Gabapentin designed for absorption throughout the intestine by high-capacity nutrient transporters. Gabapentin is an orally active P/Q type Ca2+ channel blocker. This product can be used for the study of Restless Legs Syndrome (RLS) and postherpetic neuralgia (PHN).
  • Prodrug of gabapentin.
  • Absorbed throughout the intestine by high-capacity nutrient transporters.
  • Used for the study of restless legs syndrome (RLS) and postherpetic neuralgia (PHN).
  • Stable at physiological pH.
  • Rapidly converted to gabapentin in intestinal and liver tissue from rats, dogs, monkeys, and humans.
  • Improved oral bioavailability, dose proportionality, and enhanced colonic absorption compared with gabapentin in rats and monkeys.
Catalog No. 50-003-91901