Learn More
Medchemexpress LLC Gabapentin enacarbil | 478296-72-9 | 95.0% | 329.39 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY16216
Encompass_Preferred
Gabapentin enacarbil is a prodrug of Gabapentin, designed for absorption throughout the intestine by high-capacity nutrient transporters. Gabapentin is an orally active P/Q type Ca2+ channel blocker. It can be used to study Restless Legs Syndrome (RLS) and postherpetic neuralgia (PHN).
  • Prodrug design for improved absorption.
  • Potent, orally active P/Q type Ca2+ channel blocker.
  • Research applications for restless legs syndrome (RLS) and postherpetic neuralgia (PHN).
  • Improved bioavailability in rats and monkeys.
  • Enhanced properties including increased dose proportionality and colonic absorption.
  • Stable at physiological pH but rapidly converts to gabapentin in intestinal and liver tissue.
Catalog No. 50-003-91882