Learn More
TARGETMOL CHEMICALS INC Gabapentin hydrochloride 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T6507200MG
Encompass_Preferred
Also available in 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Gabapentin hydrochloride (Neurontin HCl) is a GABA analogue, used to treat seizures and neuropathic pain. Purity 99.46%
Catalog No. 50-287-7101