Learn More
Sigma Aldrich Fine Chemicals Biosciences GABase from Pseudomonas fl
Supplier:  Sigma Aldrich Fine Chemicals Biosciences G750925UN
Encompass
GABase is a commercially available mixture composed of³-aminobutyric acid aminotransferase (GABGT) and succinic semialdehyde dehydrogenase (SSDH) obtained from Pseudomonas fluorescens.
Catalog No. 50-291-4465