Learn More
Medchemexpress LLC Gabexate mesylate | 56974-61-9 | MFCD00210299 | 98.6% | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB038510MG
Encompass_Preferred
Gabexate mesylate is a synthetic serine protease inhibitor used as a research reagent and clinically for conditions such as pancreatitis and disseminated intravascular coagulation. It inhibits trypsin-like serine proteases including thrombin, plasmin, urokinase, and Factor Xa, and is supplied as a white to off-white solid with high purity for analytical and biochemical applications.
  • Serine protease inhibitor for biochemical and cellular assays.
  • High purity suitable for analytical use.
  • White to off-white solid appearance.
  • Store sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Includes CAS number 56974-61-9 for unambiguous identification.
Catalog No. 50-002-99037