Learn More
Abcam Gabe x ate mesylate, General serine protease inhibitor, 10MG
Supplier:  Abcam AB14116710MG
Encompass_Preferred
MW 417.5 Da, Purity >99%. General serine protease inhibitor (Ki values are 3.4 nM, 5, 1.8 μM and 2.6 mM at tryptase, thrombin, tyrpsin and kallikrein serine proteases respectively). Additionally inhibits nitric oxide synthase. Displays antithrombotic activity and antitumor effect. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6842