Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gad65 (524-543) (acetate) | 98.1% | 2238.67 | 50 MG
SDP

Supplier:  Medchemexpress LLC HYP5396A50MG

Encompass_Preferred

GAD65 (524-543) acetate is a biologically active peptide comprising amino acids 524 to 543 from glutamic acid decarboxylase 65 (GAD65). This peptide is recognized for its ability to induce proliferative T cell responses in the non-obese diabetic (NOD) mouse model of spontaneous autoimmune diabetes. It specifically stimulates the diabetogenic T cell clone BDC2.5, and immunization with GAD65 (524-543) acetate can increase the susceptibility of NOD mice to type 1 diabetes induced by the adoptive transfer of BDC2.5 T cells.

  • Biologically active peptide
  • Fragment of glutamic acid decarboxylase 65 (GAD65)
  • Induces proliferative T cell responses in NOD mouse model
  • Stimulates diabetogenic T cell clone BDC2.5
  • Increases susceptibility to type 1 diabetes in NOD mice
  • Purity of 98.1% by HPLC
  • Molecular weight of 2238.67 (free base)
  • White solid appearance
  • For research use only

Catalog No. 50-003-99614


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.