Learn More
Medchemexpress LLC Gad65 (524-543) acetate | 98.1% | 2238.67 (free base) | 10 MG
Supplier: Medchemexpress LLC HYP5396A10MG
Gad65 (524-543) acetate is a biologically active peptide, a fragment of glutamic acid decarboxylase 65 (Gad65), containing amino acids 524 to 543. It induces proliferative T cell responses in non-obese diabetic (NOD) mouse models and serves as a specific stimulus for the diabetogenic T cell clone BDC2.5. Immunization with this peptide increases the susceptibility of NOD mice to type 1 diabetes.
- Biologically active peptide fragment of Gad65 (524-543)
- Induces T cell responses in non-obese diabetic (NOD) mouse models
- Stimulates diabetogenic T cell clone BDC2.5
- Increases susceptibility to type 1 diabetes in NOD mice
- Powder stable for 2 years at -80°C, 1 year at -20°C
- In solvent stable for 6 months at -80°C, 1 month at -20°C