Learn More
Cayman Chemical GAD65 MoncLNal ANTIBDYCl 100ug
SDP
Supplier:  Cayman Chemical 31360100 UG
Encompass_Preferred
Immunogen: Full-length recombinant human GAD65 • Host: Mouse • Species Reactivity: (+) Human, mouse • Cross Reactivity: None provided • Applications: ELISA, WB • MW = 65.4 kDa
Catalog No. 50-299-0934